Anti-SIRT1 Rabbit Polyclonal Antibody
Supplier: Bioss
BS-2257R
10328-870EA 1041.92 CAD
10328-870
Anti-SIRT1 Rabbit Polyclonal Antibody
Antibodies
This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class I of the sirtuin family.More Product Information