5-Carboxytetramethylrhodamine (5-TAMRA) special formulation

Supplier: Anaspec Inc

AS-81121 AS-81121-01 AS-81121-1
103010-834EA 252.31 CAD
103010-834 103010-836 103010-838
5-Carboxytetramethylrhodamine (5-TAMRA) special formulation
5-Carboxytetramethylrhodamine (5-TAMRA)

5-TAMRA is the purified single isomer of 5(6)-TAMRA. It is preferred for some complicated biological applications where reproducibility is more critical than material cost since the minor positional difference between 5-TAMRA and 6-TAMRA might affect some biological properties of the underlying conjugates.


  • Spectral properties: Abs/Em = 541/568 nm
  • Solvent system: DMF or DMSO
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice

Formula: C₂₅H₂₂N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD00269769
CAS Number: 91809-66-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR