Histone H4 (8-30)-WGK

Supplier: Anaspec Inc

AS-65417-1
103009-604EA 869.23 CAD
103009-604
Histone H4 (8-30)-WGK
Proteins and Peptides

This peptide is Histone H4 amino acid residues 8-30 with a C-terminal WG linker followed by a biotinylated lysine.
Sequence:Ac-KGLGKGGAKRHRKVLRDNIQGIT-WGK(biotin)
MW:3142.7 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR