[Arg(Me2s)26]-Histone H3 (15-36)-GGK,Biotin

Supplier: Anaspec Inc

AS-65416-1
103009-600EA 949.23 CAD
103009-600
[Arg(Me2s)26]-Histone H3 (15-36)-GGK,Biotin
Proteins and Peptides

This peptide is Histone H3 amino acid residues 15-36 with a C-terminal GG linker followed by a biotinylated lysine. It is symmetrically dimethylated at arginine 26 with a methyl group added to each nitrogen of the guanidinium group.
Sequence:APRKQLATKAA-R(Me2s)-KSAPATGGVK-GGK(biotin)
MW:2704.2 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR