FN-A208 Fusion Peptide

Supplier: Anaspec Inc

AS-65007
103008-646EA 267.69 CAD
103008-646
FN-A208 Fusion Peptide
Proteins and Peptides

This peptide is a fusion of A208, derived from murine laminin a1, and the active site of fibronectin (GRGDS), with a glycine spacer. This peptide forms amyloid-like fibrils and promotes formation of actin stress fibers that mediate fibroblast cell attachment, offering it potential as a bioadhesive for tissue regeneration and engineering. FN-A208 interacts with IKVAV receptors and integrins. Its activity is disrupted by the presence of EDTA.
Sequence:GRGDSGAASIKVAVSADR
MW:1716.9 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR