[Lys(Me1)9]-Histone H3 (1-21)-GGK,Biotin

Supplier: Anaspec Inc

AS-64626-1
103008-346EA 869.23 CAD
103008-346
[Lys(Me1)9]-Histone H3 (1-21)-GGK,Biotin
Proteins and Peptides

This peptide is Histone H3 amino acid residues 1-21. It is monomethylated at lysine 9 with a C-terminal GG linker followed by a biotinylated lysine. The monomethylation of histone H3 at lysine 9 is dynamically and sex-differentially regulated during meiotic prophase. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:ARTKQTAR-K(Me1)-STGGKAPRKQLA-GGK(Biotin)-NH2
MW:2736.2 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR