Human Beta-Amyloid (1-10)

Supplier: Anaspec Inc

AS-64478
103008-188EA 172.31 CAD
103008-188
Human Beta-Amyloid (1-10)
Proteins and Peptides

This peptide is beta-Amyloid (1-10), human sequence.
Sequence: DAEFRHDSGY
Molecular Weight: 1196.2 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR