[Lys(Me3)36]-Histone H3 (21-44)-GK,Biotin

Supplier: Anaspec Inc

AS-64441-025 AS-64441-1
103008-162EA 310.77 CAD
103008-162 103008-164
[Lys(Me3)36]-Histone H3 (21-44)-GK,Biotin
Proteins and Peptides

This is a Histone 3 peptide trimethylated at lysine 36. It's role has been indicative of defining exons, as certain nucleosome-enriched exons are observed to be enriched in some histone modifications. It is belived that this pattern of modification as seen with this H3K36me3 peptide, influences alternative splicing, perhaps by signaling effector proteins to mark particular exons for inclusion in the final transcript as they exit the RNA Polymerase II complex. It has also been shown in yeast that H3K36me3 gets deposited on histones as they are displaced by RNA polymerase II (RNAPII) during transcription. H3K36me3 then serves as a mark for HDACs to bind and deacetylate the histones.
Sequence:ATKAARKSAPATGGV-K(Me3)-KPHRYRPG-GK(Biotin)
MW:2959.5 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR