Histone H3 (1-21), K4 Methylated, FAM (Carboxyfluorescein)

Supplier: Anaspec Inc

AS-63825
103007-758EA 941.54 CAD
103007-758
Histone H3 (1-21), K4 Methylated, FAM (Carboxyfluorescein)
Proteins and Peptides

This is a FAM (Abs/Em = 492/518 nm) labeled histone 3 (H3) amino acid residues 1 to 21 with lysine 4 methylated.
Sequence:ART-K(Me1)-QTARKSTGGKAPRKQLA-GGK(FAM)-NH2
MW:2868.2 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR