Human;Mouse;Rat Beta-Amyloid (23-42)

Supplier: Anaspec Inc

AS-62766
103007-516EA 356.92 CAD
103007-516
Human;Mouse;Rat Beta-Amyloid (23-42)
Proteins and Peptides

This is amino acids 23 to 42 fragment of beta-amyloid.
Sequence: DVGSNKGAIIGLMVGGVVIA
Molecular Weight: 1870.3 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR