S. pneumoniae CFP10 (71–85)

Supplier: Anaspec Inc

AS-61689
103006-902EA 252.31 CAD
103006-902
S. pneumoniae CFP10 (71–85)
Proteins and Peptides

This 15-mer peptide is fragment of (CFP)10 protein, which is secreted from mycobacterium tuberculosis 10-kDa culture filtrate stimulate IFN-gamma; production and CTL activity by CD4+ and CD8+ cells, from persons expressing a spectrum of MHC molecules. This peptide is an excellent candidate for inclusion in a subunit antituberculosis vaccine.
Sequence:EISTNIRQAGVQYSR
MW:1721.9 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now

SPECIFICATIONS

Protein/peptide name CFP10 (71–85)
Environmentally Preferable
Protein synonyms CFP10 Peptide
Species S. pneumoniae
Source
Conjugation Unconjugated
Sequence EISTNIRQAGVQYSR
Purity 95%
Molecular weight 1721.9 Da
Storage conditions –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR