Human Papillomavirus (HPV) E7 protein (49-57)

Supplier: Anaspec Inc

AS-61022
103006-258EA 172.31 CAD
103006-258
Human Papillomavirus (HPV) E7 protein (49-57)
Proteins and Peptides

This peptide is a H-2Db-restricted epitope from human Papillomavirus E7 protein (49-57).
Sequence:RAHYNIVTF
MW:1120.3 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR