HIV TAT (47-57) Peptide

Supplier: Anaspec Inc

AS-60023-1 AS-60023-5
103005-824EA 252.31 CAD
103005-824 103005-826
HIV TAT (47-57) Peptide
Proteins and Peptides

This is the most characterized fragment of the HIV transactivator protein (TAT). This arginine-rich TAT peptide penetrates plasma membrane directly, but not through endocytosis.
Sequence: YGRKKRRQRRR
MW: 1559.9 Da
% Peak area by HPLC: 95
Storage condition: -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR