SAMS Peptide

Supplier: Anaspec Inc

AS-60167-5
103002-738EA 1190.77 CAD
103002-738
SAMS Peptide
Proteins and Peptides

A synthetic peptide substrate specific for AMP-activated protein kinase (AMPK).
Sequence:HMRSAMSGLHLVKRR-NH2
MW:1778.2 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR