[Arg8]-Vasopressin (AVP) Peptide

Supplier: Anaspec Inc

AS-24289 AS-24290
102996-516EA 106.15 CAD
102996-516 102996-518
[Arg8]-Vasopressin (AVP) Peptide
Proteins and Peptides

AVP have been identified as an important regulator of fluid and electrolyte homeostasis through its anti-diuretic action on the kidney, After myocardial infarction, plasma levels of [Arg8]-vasopressin rise to recover hemodynamics.
Sequence: CYFQNCPRG-NH2 (Disulfide bridge: 1-6)
MW: 1084.3 Da
% Peak area by HPLC: 95
Storage condition: -20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR