[Succinyl-Asp6, NMePhe8]-Substance P (6-11)

Supplier: Anaspec Inc

AS-22887
102996-324EA 503.08 CAD
102996-324
[Succinyl-Asp6, NMePhe8]-Substance P (6-11)
Proteins and Peptides

Senktide is a NK3 tachykinin receptor agonist. It was shown to induce bronchoconstriction in guinea pig lungs.
Sequence:Suc-DF-(NMeF)-GLM-NH2
MW:842.1 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now

SPECIFICATIONS

Protein/peptide name [Succinyl-Asp6, NMePhe8]-Substance P (6-11)
Protein synonyms Substance P|NK3 tachykinin receptor agonist Peptide|Senktide
Environmentally Preferable
Species
Source
Conjugation Unconjugated
Sequence Suc-DF-(NMeF)-GLM-NH2
Purity 95%
Molecular weight 842.1 Da
Storage conditions –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR