Anti-GARP Monoclonal Antibody [clone: ANC8C9]

Supplier: Adipogen
ANC-277-020
102979-308EA 484.62 CAD
102979-308
Anti-GARP Monoclonal Antibody [clone: ANC8C9]
Antibodies
GARP (LRRC32) is a ~80kD transmembrane glycoprotein expressed on the cell surface of megakaryocytes, platelets and a subset of activated regulatory T (Treg) cells. It is essential for the surface expression of latent TGF-beta on platelets and activated FOXP3+ regulatory T cells.

Type: Primary
Antigen: GARP
Clonality: Monoclonal
Clone: ANC8C9
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2akappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR