Anti-GIP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-0098R-FITC
10230-896EA 1300.77 CAD
10230-896
Anti-GIP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Potent stimulator of insulin secretion and relatively poor inhibitor of gastric acid secretion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GIP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR