Anti-CASP9 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-0049R-CY5.5
10229-972EA 1300.77 CAD
10229-972
Anti-CASP9 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Involved in the activation cascade of caspases responsible for apoptosis execution. Binding of caspase-9 to Apaf-1 leads to activation of the protease which then cleaves and activates caspase-3. Promotes DNA damage-induced apoptosis in a ABL1/c-Abl-dependent manner. Proteolytically cleaves poly(ADP-ribose) polymerase (PARP). Isoform 2 lacks activity is an dominant-negative inhibitor of caspase-9.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR