Fluorescent Wash Buffer, Azure Biosystems
Supplier: Azure Biosystems
AC2145
10147-340EA 420 CAD
10147-340
Fluorescent Wash Buffer, Azure Biosystems
Electrophoresis Reagents Western Blotting Reagents
Azure IR Fluorescent Blot Washing Buffer is optimized for use with visible and infrared fluorescent Western blotting.
  • Optimized for use with visible and infrared fluorescent Western blotting
  • Provided as a 10X concentrate
Azure IR Fluorescent Blot Washing Buffer is optimized for use with visible and infrared fluorescent Western blotting. Provided as a 10X concentrate.
Ordering information: 10X Fluorescent Blot Washing Buffer