Anti-PTCHD3 Goat Polyclonal Antibody

Supplier: Prosci
43-006
10115-186EA 1131.25 CAD
10115-186
Anti-PTCHD3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. The starting concentration recommended for Western Blot is 0.3-1?g/mL.

PTCHD3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PTCHD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR