Anti-CRTC2 Goat Polyclonal Antibody

Supplier: Prosci
42-852
10115-014EA 1131.25 CAD
10115-014
Anti-CRTC2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 75 kDa band observed in mouse and rat brain and spinal cord lysates (calculated MW of 73.3 kDa according to mouse NP_859066.1 and 73.1 kDa according to rat NP_001029067.1). Recommended concentration: 0.03-0.1 ?g/mL.

CRTC2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CRTC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR