Anti-IGF1 Goat Polyclonal Antibody

Supplier: Prosci
42-659
10114-740EA 1131.25 CAD
10114-740
Anti-IGF1 Goat Polyclonal Antibody
Antibodies
Approximately 15 kDa band observed in human uterus lysates (calculated MW of 15.2 kDa according to NP_001104754.1). Recommended concentration 0.5-2 ?g/mL.

IGF1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IGF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR