Anti-ETFDH Goat Polyclonal Antibody

Supplier: Prosci
42-621
10114-692EA 1131.25 CAD
10114-692
Anti-ETFDH Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Approximately 70 kDa band observed in human, mouse and rat heart lysates (calculated MW of 68.5 kDa according to NP_004444.2). Recommended concentration: 1-3 ?g/mL.

ETFDH antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ETFDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol ETFDH
Antigen name electron-transferring-flavoprotein dehydrogenase
Environmentally Preferable
Antigen synonyms ETFDH
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Goat
Gene ID Q16134
Isotype
Western blot Yes
ELISA Yes
Immunogen ETFDH antibody was raised against a 12 amino acid synthetic peptide near the Internal region of ETFDH.
Form Liquid
Purification ETFDH antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Concentration 500 ug/mL
Storage buffer ETFDH antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
Storage temperature Aliquot and store at –20 °C. Minimize freezing and thawing.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR