Anti-CST8 Goat Polyclonal Antibody

Supplier: Prosci
42-303
10113-760EA 1131.25 CAD
10113-760
Anti-CST8 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 37 kDa band in human testis lysates after 1 ?g/mL antibody staining. The 37 kDa band was successfully blocked by incubation with the immunizing peptide.

CST8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CST8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol CST8
Antigen name cystatin 8 (cystatin-related epididymal specific)
Environmentally Preferable
Antigen synonyms CST8
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Goat
Gene ID O60676
Isotype
Western blot Yes
ELISA Yes
Immunogen CST8 antibody was raised against a 12 amino acid synthetic peptide near the internal region of CST8.
Form Liquid
Purification CST8 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Concentration 500 ug/mL
Storage buffer CST8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
Storage temperature Aliquot and store at –20 °C. Minimize freezing and thawing.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR