Anti-AIRE Goat Polyclonal Antibody

Supplier: Prosci
45-226
10113-206EA 1131.25 CAD
10113-206
Anti-AIRE Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Appr. 60 kDa in transfected HEK293 lysates (calculated MW of 57.7 kDa according to NP_000374.1). Recommended concentration: 0.5-1.0 ?g/mL. Immunohistochemistry: In paraffin embedded human thymus shows nuclear staining in select thymocytes. Recommended concentration, 2.5 ?g/mL.

AIRE antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AIRE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR