Anti-GABRP Rabbit Polyclonal Antibody

Supplier: Prosci
31-108
10111-174EA 1097.39 CAD
10111-174
Anti-GABRP Rabbit Polyclonal Antibody
Antibodies
The gamma-aminobutyric acid (GABA) A receptor is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. The subunit encoded by this gene is expressed in several non-neuronal tissues including the uterus and ovaries. This subunit can assemble with known GABA A receptor subunits, and the presence of this subunit alters the sensitivity of recombinant receptors to modulatory agents such as pregnanolone.

GABRP antibody can be used for detection of GABRP by ELISA at 1:1562500. GABRP antibody can be used for detection of GABRP byMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol GABRP
Environmentally Preferable
Antigen name gamma-aminobutyric acid (GABA) A receptor, pi
Antigen synonyms GABRP
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Dog,Rat,Mouse
Host Rabbit
Gene ID O00591
Isotype
Western blot Yes
ELISA Yes
Immunogen Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human GABRP.
Form Liquid
Molecular weight 51 kDa
Storage buffer Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Storage temperature For short periods of storage (days) store at 4 °C. For longer periods of storage, store GABRP antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR