Anti-ARHGAP28 Rabbit Polyclonal Antibody
Supplier: Prosci
30-936
10110-944EA 1202.19 CAD
10110-944
Anti-ARHGAP28 Rabbit Polyclonal Antibody
Antibodies
ARHGAP28 contains 1 Rho-GAP domain. ARHGAP28 is a GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
ARHGAP28 antibody can be used for detection of ARHGAP28 by ELISA at 1:1562500. ARHGAP28 antibody can be used for detection of ARHGAP28 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot andMore Product Information