Anti-ASPH Rabbit Polyclonal Antibody
Supplier: Prosci
30-725
10110-586EA 1097.39 CAD
10110-586
Anti-ASPH Rabbit Polyclonal Antibody
Antibodies
ASPH is thought to play an important role in calcium homeostasis. Alternative splicing of this gene results in five transcript variants which vary in protein translation, the coding of catalytic domains, and tissue expression. Variation among these transcripts impacts their functions which involve roles in the calcium storage and release process in the endoplasmic and sarcoplasmic reticulum as well as hydroxylation of aspartic acid and asparagine in epidermal growth factor-like domains of various proteins.This gene is thought to play an important role in calcium homeostasis. AlternativeMore Product Information