Anti-ZSCAN5C Rabbit Polyclonal Antibody
Supplier: Prosci
30-683
10110-466EA 1202.19 CAD
10110-466
Anti-ZSCAN5C Rabbit Polyclonal Antibody
Antibodies
The exact function of LOC649137 remains unknown.
LOC649137 antibody can be used for detection of LOC649137 by ELISA at 1:1562500. LOC649137 antibody can be used for detection of LOC649137 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: ZSCAN5C
More Product Information