Anti-SPATA12 Rabbit Polyclonal Antibody
Supplier: Prosci
30-670
10110-444EA 1202.19 CAD
10110-444
Anti-SPATA12 Rabbit Polyclonal Antibody
Antibodies
Putative function of SPATA12 is to maintain the cell in a differentiated state and/or to suppress cell proliferation in the human testis.
SPATA12 antibody can be used for detection of SPATA12 by ELISA at 1:312500. SPATA12 antibody can be used for detection of SPATA12 by western blot at 0.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -2More Product Information