Anti-PRKCZ Rabbit Polyclonal Antibody
Supplier: Prosci
29-814
10109-010EA 1202.19 CAD
10109-010
Anti-PRKCZ Rabbit Polyclonal Antibody
Antibodies
Protein kinase C (PKC) zeta is a member of the PKC family of serine/threonine kinases which are involved in a variety of cellular processes such as proliferation, differentiation and secretion. Unlike the classical PKC isoenzymes which are calcium-dependent, PKC zeta exhibits a kinase activity which is independent of calcium and diacylglycerol but not of phosphatidylserine. Furthermore, it is insensitive to typical PKC inhibitors and cannot be activated by phorbol ester. Unlike the classical PKC isoenzymes, it has only a single zinc finger module. These structural and biochemical propertiesMore Product Information