Anti-ZNF496 Rabbit Polyclonal Antibody
Supplier: Prosci
29-113
10107-628EA 1097.39 CAD
10107-628
Anti-ZNF496 Rabbit Polyclonal Antibody
Antibodies
ZNF496 contains a SCAN box, a KRAB-A domain, and four consensus C2H2-type zinc fingers preceded by a unique finger derivative, referred to herein as the C2HR motif. The C2HR motif functions to mediate protein-protein interaction with the cysteine-rich (C5HCH) domain of NSD1 in a Zn (II)-dependent fashion, and when tethered to RNA polymerase II promoters, represses transcription in an NSD1-dependent manner. ZNF496 contains a novel type of zinc finger motif that functions as a docking site for NSD1 and is more than just a degenerate evolutionary remnant of a C2H2 motif.
ZNF496 antibody canMore Product Information