Anti-SOX12 Rabbit Polyclonal Antibody
Supplier: Prosci
27-832
10105-532EA 1097.39 CAD
10105-532
Anti-SOX12 Rabbit Polyclonal Antibody
Antibodies
Members of the SOX family of transcription factors are characterized by the presence of a DNA-binding high mobility group (HMG) domain, homologous to the HMG box of sex-determining region Y (SRY). Forming a subgroup of the HMG domain superfamily, SOX proteins have been implicated in cell fate decisions in a diverse range of developmental processes. SOX transcription factors have diverse tissue-specific expression patterns during early development and have been proposed to act as target-specific transcription factors and/or as chromatin structure regulatory elements. The protein encoded byMore Product Information