Anti-FDXR Rabbit Polyclonal Antibody
Supplier: Prosci
26-915
10104-192EA 1201.89 CAD
10104-192
Anti-FDXR Rabbit Polyclonal Antibody
Antibodies
FDXR is a mitochondrial flavoprotein that initiates electron transport for cytochromes P450 receiving electrons from NADPH.This gene encodes a mitochondrial flavoprotein that initiates electron transport for cytochromes P450 receiving electrons from NADPH. Multiple alternatively spliced transcript variants of this gene have been described although the full-length nature of only two that encode different isoforms have been determined.
FDXR antibody can be used for detection of FDXR by ELISA at 1:12500. FDXR antibody can be used for detection of FDXR by western blot at 1 ?g/mL, and HRPMore Product Information