Anti-TCTE1 Rabbit Polyclonal Antibody
Supplier: Prosci
26-791
10104-096EA 1202.19 CAD
10104-096
Anti-TCTE1 Rabbit Polyclonal Antibody
Antibodies
TCTE1 contains 7 LRR (leucine-rich) repeats. The exact function of TCTE1 remains unknown.
TCTE1 antibody can be used for detection of TCTE1 by ELISA at 1:312500. TCTE1 antibody can be used for detection of TCTE1 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.