Anti-DPH2 Rabbit Polyclonal Antibody
Supplier: Prosci
26-626
10103-894EA 1202.19 CAD
10103-894
Anti-DPH2 Rabbit Polyclonal Antibody
Antibodies
DPH2 gene is one of two human genes similar to the yeast gene dph2. The yeast gene was identified by its ability to complement a diphthamide mutant strain, and thus probably functions in diphthamide biosynthesis. Diphthamide is a post-translationally modified histidine residue present in elongation factor 2 (EF2) that is the target of diphtheria toxin ADP-ribosylation. Two transcript variants encoding different isoforms have been found for this gene.This gene is one of two human genes similar to the yeast gene dph2. The yeast gene was identified by its ability to complement a diphthamideMore Product Information