Anti-GPHN Rabbit Polyclonal Antibody
Supplier: Prosci
26-621
10103-884EA 1202.19 CAD
10103-884
Anti-GPHN Rabbit Polyclonal Antibody
Antibodies
GPHN is a neuronal assembly protein that anchors inhibitory neurotransmitter receptors to the postsynaptic cytoskeleton via high affinity binding to a receptor subunit domain and tubulin dimers. In nonneuronal tissues, the protein is also required for molybdenum cofactor biosynthesis. Mutations in this gene may be associated with the neurological condition hyperplexia and also lead to molybdenum cofactor deficiency. Numerous alternatively spliced transcript variants encoding different isoforms have been described; however, the full-length nature of all transcript variants is not currentlyMore Product Information