Anti-OAT Rabbit Polyclonal Antibody
Supplier: Prosci
26-283
10103-386EA 1202.19 CAD
10103-386
Anti-OAT Rabbit Polyclonal Antibody
Antibodies
OAT is a key enzyme in the pathway that converts arginine and ornithine into the major excitatory and inhibitory neurotransmitters glutamate and GABA. Mutations of this enzyme cause the autosomal recessive eye disease Gyrate Atrophy. This gene encodes the mitochondrial enzyme ornithine aminotransferase, which is a key enzyme is the pathway that converts arginine and ornithine into the major excitatory and inhibitory neurotransmitters glutamate and GABA. Mutations that result in a deficiency of this enzyme cause the autosomal recessive eye disease Gyrate Atrophy. Publication Note: This RefSeqMore Product Information