Anti-CREBBP Rabbit Polyclonal Antibody
Supplier: Prosci
25-879
10102-796EA 1202.19 CAD
10102-796
Anti-CREBBP Rabbit Polyclonal Antibody
Antibodies
CREBBP is involved in the transcriptional coactivation of many different transcription factors. First isolated as a nuclear protein that binds to cAMP-response element binding protein (CREB), this gene is now known to play critical roles in embryonic development, growth control, and homeostasis by coupling chromatin remodeling to transcription factor recognition. CREBBP has intrinsic histone acetyltransferase activity and also acts as a scaffold to stabilize additional protein interactions with the transcription complex. This protein acetylates both histone and non-histone proteins. ThisMore Product Information