Anti-MAOB Rabbit Polyclonal Antibody
Supplier: Prosci
25-875
10102-788EA 1201.89 CAD
10102-788
Anti-MAOB Rabbit Polyclonal Antibody
Antibodies
MAOB belongs to the flavin monoamine oxidase family. It is a enzyme located in the mitochondrial outer membrane. It catalyzes the oxidative deamination of biogenic and xenobiotic amines and plays an important role in the metabolism of neuroactive and vasoactive amines in the central nervous sysytem and peripheral tissues. This protein preferentially degrades benzylamine and phenylethylamine. The protein encoded by this gene belongs to the flavin monoamine oxidase family. It is a enzyme located in the mitochondrial outer membrane. It catalyzes the oxidative deamination of biogenic andMore Product Information