Anti-CLEC4M Rabbit Polyclonal Antibody

Supplier: Prosci
25-765
10102-592EA 1202.19 CAD
10102-592
Anti-CLEC4M Rabbit Polyclonal Antibody
Antibodies
CLEC4M is a transmembrane receptor and is often referred to as L-SIGN because of its expression in the endothelial cells of the lymph nodes and liver. It is involved in the innate immune system and recognizes numerous evolutionarily divergent pathogens ranging from parasites to viruses, with a large impact on public health. The protein is organized into three distinct domains: an N-terminal transmembrane domain, a tandem-repeat neck domain and C-type lectin carbohydrate recognition domain. The extracellular region consisting of the C-type lectin and neck domains has a dual function as aMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CLEC4M
Antigen name C-type lectin domain family 4, member M
Environmentally Preferable
Antigen synonyms CLEC4M
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID Q9H2X3
Isotype
Western blot Yes
ELISA Yes
Immunogen Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human CLEC4M.
Form Liquid
Molecular weight 30 kDa
Purification Antibody is purified by peptide affinity chromatography method.
Storage buffer Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Storage temperature For short periods of storage (days) store at 4 °C. For longer periods of storage, store CLEC4M antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR