Anti-IFIT2 Rabbit Polyclonal Antibody
Supplier: Prosci
25-735
10102-532EA 1202.19 CAD
10102-532
Anti-IFIT2 Rabbit Polyclonal Antibody
Antibodies
IFIT2 belongs to the IFIT family. It contains 6 TPR repeats. IFIT2 inhibits migration activity and increases survival of oral squamous cell carcinoma.
IFIT2 antibody can be used for detection of IFIT2 by ELISA at 1:62500. IFIT2 antibody can be used for detection of IFIT2 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store atMore Product Information