Anti-ZNF555 Rabbit Polyclonal Antibody

Supplier: Prosci
28-455
10101-314EA 1202.19 CAD
10101-314
Anti-ZNF555 Rabbit Polyclonal Antibody
Antibodies
ZNF555 contains 15 C2H2-type zinc fingers and 1 KRAB domain. It belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation. Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.

ZNF555 antibody can be used for detection of ZNF555 by ELISA at 1:12500. ZNF555 antibody can be used for detection of ZNF555 by western blot at 0.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptideMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ZNF555
Antigen name zinc finger protein 555
Environmentally Preferable
Antigen synonyms ZNF555
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID Q8NEP9
Isotype
Western blot Yes
ELISA Yes
Immunogen Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human ZNF555.
Form Liquid
Molecular weight 73 kDa
Purification Antibody is purified by peptide affinity chromatography method.
Storage buffer Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Storage temperature For short periods of storage (days) store at 4 °C. For longer periods of storage, store ZNF555 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR