Anti-CDK5 Rabbit Polyclonal Antibody
Supplier: Prosci
27-214
10100-612EA 1202.19 CAD
10100-612
Anti-CDK5 Rabbit Polyclonal Antibody
Antibodies
CDK5 is probably involved in the control of the cell cycle. It interacts with D1 and D3-type G1 cyclins. The protein can phosphorylate histone H1, tau, MAP2 and NF-H and NF-M. And it is also thought to interacts with p35 which activates the kinase.
CDK5 antibody can be used for detection of CDK5 by ELISA at 1:62500. CDK5 antibody can be used for detection of CDK5 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose.More Product Information