Anti-FUCA1 Rabbit Polyclonal Antibody
Supplier: Prosci
26-844
10100-322EA 1201.89 CAD
10100-322
Anti-FUCA1 Rabbit Polyclonal Antibody
Antibodies
Alpha-L-fucosidase (EC 3.2.1.51) is a lysosomal enzyme involved in the degradation of fucose-containing glycoproteins and glycolipids. At least 2 separate polymorphic alpha-L-fucosidases are recognized in man: that in tissues, FUCA1, which is deficient in fucosidosis, and that in plasma, FUCA2. Fucosidosis is an autosomal recessive lysosomal storage disease caused by defective alpha-L-fucosidase with accumulation of fucose in the tissues. Different phenotypes include clinical features such as neurologic deterioration, growth retardation, visceromegaly, and seizures in a severe early form;More Product Information