Human Recombinant IL23 (from (BTI-Tn-5B1-4) Hi-5 Insect Cells)
Supplier: Prosci
40-559
10073-030EA 481.9 CAD
10073-030
Human Recombinant IL23 (from (BTI-Tn-5B1-4) Hi-5 Insect Cells)
Proteins and Peptides
IL-23 is a proinflammatory heterodimeric protein composed of two subunits, a unique p19 subunit and a p40 subunit, which is shared with IL-12. IL-23 is secreted by activated dendritic cells and macrophages, and signals though a receptor comprised of IL-23R complexed with IL-12Rβ2. IL-23 has been shown to enhance proliferation of memory T cells. It also stimulates the production of IFN-gamma in NK cells, induces IL-17 production, and drives Th17 mediated responses. Recombinant IL-23 is a 53.5 kDa heterodimeric protein consisting of two subunits, p19 (170 amino acids) and p40 (306 amino acids).*Manufactured using BTI-Tn-5B1-4 cells under license from the Boyce Thompson Institute for Plant Research, Inc.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.