Anti-FBXL3 Rabbit Polyclonal Antibody
Supplier: Prosci
79-664
10071-928EA 866.01 CAD
10071-928
Anti-FBXL3 Rabbit Polyclonal Antibody
Antibodies
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, inMore Product Information