Anti-UNG Rabbit Polyclonal Antibody
Supplier: Prosci
7353
10061-896EA 917.13 CAD
10061-896
Anti-UNG Rabbit Polyclonal Antibody
Antibodies
UNG2 Antibody: The human uracil-DNA glycosylase (UNG) gene encodes both mitochondrial (UNG1) and nuclear (UNG2) forms through differentially regulated promoters and alternative splicing. UNG2 is the major enzyme in the base excision repair pathway that removes uracil residues from DNA that arise through either misincorporation during replication or cytosine deamination. UNG2 can also be bound by the HIV-1 integrase and incorporated into the virion particle, suggesting that it is required to remove uracils from the viral genome. As the intrinsic antiviral protein APOBEC3G generates numerousMore Product Information