Anti-PRRT2 Rabbit Polyclonal Antibody
Supplier: Prosci
7295
10061-866EA 917.13 CAD
10061-866
Anti-PRRT2 Rabbit Polyclonal Antibody
Antibodies
PRRT2 Antibody: The proline-rich transmembrane protein 2 (PRRT2) contains a proline-rich domain in its N-terminal half and is predominantly expressed in brain and spinal cord in embryonic and postnatal stages. While little is known of the function of this protein, mutations in PRRT2 have been shown to be the causative gene of paroxysmal kinesigenic dyskinesia, which is characterized by recurrent, brief attacks of abnormal involuntary movements induced by sudden voluntary movements. Recent studies have shown that PRRT2 may also be involved in some forms of benign familial infantile epilepsy (More Product Information